Learn More
Glass Expansion GazFit Connectors, 2 for 6mm OD side arms, 2 for 4mm OD side arms (PKT.4)
SDP
Catalog No. 502702980
Encompass_Preferred
GazFit Connectors. Package of 4 GazFit Connectors that includes 2 for 4mm OD side arms, 2 for 6mm OD side arms. Typically used with torch argon side arms and laser ablation adaptors to connect soft walled tubing.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2980 Supplier Glass Expansion Supplier No. GAZ0604
GazFit Connectors. Package of 4 GazFit Connectors that includes 2 for 4mm OD side arms, 2 for 6mm OD side arms. Typically used with torch argon side arms and laser ablation adaptors to connect soft walled tubing.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001