Learn More
Glass Expansion GazFit Connectors for 5mm OD torch side arm (PKT 4)
SDP
Catalog No. 502702983
Encompass_Preferred
GazFit Connectors for 5mm OD torch side arms (PKT 4). Compatible with 5mm OD torch side arms. Makes connection to torch side arms with soft walled tubing for argon gas supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2983 Supplier Glass Expansion Supplier No. GAZ05
GazFit Connectors for 5mm OD torch side arms (PKT 4). Compatible with 5mm OD torch side arms. Makes connection to torch side arms with soft walled tubing for argon gas supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001