Learn More
Glass Expansion GazFit Connectors for 6mm OD side arm (PKT.4)
SDP
Catalog No. 502702982
Encompass_Preferred
GazFit Connectors for 6mm OD torch side arms (PKT 4). Compatible with PerkinElmer Elan 6000, 9000, DRC II, DRC-E and NexION 300/350 ICP-MS Standard Quartz Torch (30-808-8107) and High Efficiency Quartz Torch (501907145, 31-808-2784). Makes connection to torch side arms with soft walled tubing for argon gas supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2982 Supplier Glass Expansion Supplier No. GAZ06
GazFit Connectors for 6mm OD torch side arms (PKT 4). Compatible with PerkinElmer Elan 6000, 9000, DRC II, DRC-E and NexION 300/350 ICP-MS Standard Quartz Torch (30-808-8107) and High Efficiency Quartz Torch (501907145, 31-808-2784). Makes connection to torch side arms with soft walled tubing for argon gas supply.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001