Learn More
Glass Expansion GazFit Connectors for 6mm OD side arm with connection for 1/8inch ID tubing (PKT.4)
SDP
Catalog No. 502702981
Encompass_Preferred
GazFit Connectors. Package of 4 GazFit Connectors for 6mm OD side arm with connection for 3.2mm (1/8”) ID tubing.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2981 Supplier Glass Expansion Supplier No. GAZ063.2B
GazFit Connectors. Package of 4 GazFit Connectors for 6mm OD side arm with connection for 3.2mm (1/8”) ID tubing.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001