Learn More
Glass Expansion GazFit Union 4mm (PKT 2)
SDP
Catalog No. 502702984
Encompass_Preferred
GazFit Union 4mm (PKT 2). GazFit Union Connector for 4mm OD laser ablation adaptors and 4mm OD rigid walled tubing. Compatible with the following laser ablation adaptors: 31-808-3863, 21-809-4140, and 31-808-3045.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2984 Supplier Glass Expansion Supplier No. GAZ04U
Add to Cart
Add to Cart
GazFit Union 4mm (PKT 2). GazFit Union Connector for 4mm OD laser ablation adaptors and 4mm OD rigid walled tubing. Compatible with the following laser ablation adaptors: 31-808-3863, 21-809-4140, and 31-808-3045.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001