Learn More
Medchemexpress LLC GB-110 hydrochloride | 99.86% | 645.23 | 50 MG
SDP
Catalog No. 5000436053
Encompass_Preferred
GB-110 hydrochloride is a potent, orally active, and nonpeptidic protease activated receptor 2 (PAR2) agonist. It selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM.
  • Potent
  • Orally active
  • Nonpeptidic protease activated receptor 2 (PAR2) agonist
  • Selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM
Catalog No. 50-004-36053 Supplier Medchemexpress LLC Supplier No. HY120528A50MG
GB-110 hydrochloride is a potent, orally active, and nonpeptidic protease activated receptor 2 (PAR2) agonist. It selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM.
  • Potent
  • Orally active
  • Nonpeptidic protease activated receptor 2 (PAR2) agonist
  • Selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM