Learn More
Medchemexpress LLC GB-110 hydrochloride | 99.9% | 645.23 | 1 ML
SDP
Catalog No. 5000436784
Encompass_Preferred
GB-110 hydrochloride is a potent, orally active, and nonpeptidic protease activated receptor 2 (PAR2) agonist. It selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM.
  • Potent, orally active, and nonpeptidic PAR2 agonist.
  • Selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells.
  • Exhibits an EC50 of 0.28 μM for PAR2-mediated intracellular Ca2+ release in HT29 cells.
  • Equipotent with 2f-LIGRLO-NH2 and 10-fold more potent than SLIGRL-NH2 in iCa2+ mobilization assay in HT29 cells.
Catalog No. 50-004-36784 Supplier Medchemexpress LLC Supplier No. HY120528A10MM/1ML
GB-110 hydrochloride is a potent, orally active, and nonpeptidic protease activated receptor 2 (PAR2) agonist. It selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM.
  • Potent, orally active, and nonpeptidic PAR2 agonist.
  • Selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells.
  • Exhibits an EC50 of 0.28 μM for PAR2-mediated intracellular Ca2+ release in HT29 cells.
  • Equipotent with 2f-LIGRLO-NH2 and 10-fold more potent than SLIGRL-NH2 in iCa2+ mobilization assay in HT29 cells.