Learn More
Medchemexpress LLC GB-110 hydrochloride | 99.9% | 645.23 | 100 MG
SDP
Catalog No. 5000437165
Encompass_Preferred
GB-110 hydrochloride is a potent, orally active, and nonpeptidic protease activated receptor 2 (PAR2) agonist. It selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM.
  • Potent and orally active
  • Nonpeptidic compound
  • Selective PAR2 agonist
  • Induces Ca2+ release in HT29 cells
  • Intended for research use only
  • High purity
Catalog No. 50-004-37165 Supplier Medchemexpress LLC Supplier No. HY120528A100MG
GB-110 hydrochloride is a potent, orally active, and nonpeptidic protease activated receptor 2 (PAR2) agonist. It selectively induces PAR2-mediated intracellular Ca2+ release in HT29 cells with an EC50 of 0.28 μM.
  • Potent and orally active
  • Nonpeptidic compound
  • Selective PAR2 agonist
  • Induces Ca2+ release in HT29 cells
  • Intended for research use only
  • High purity