Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gb-88 | 1416435-96-5 | 98.7% | 25 MG
SDP

Catalog No. 5000232944
Encompass_Preferred

GB-88 is an oral, selective non-peptide antagonist of protease-activated receptor 2 (PAR2) that inhibits PAR2-activated intracellular Ca2+ release with an IC50 of 2 μM. It is supplied as a research reagent, typically provided in milligram-scale packs and commonly dissolved in DMSO for assays.

  • Oral, selective non-peptide PAR2 antagonist.
  • Inhibits PAR2-activated intracellular Ca2+ release (IC50 = 2 μM).
  • High purity suitable for pharmacology studies (98.7%).
  • Supplied in milligram pack sizes for research use.
  • Soluble in DMSO for assay preparation.
  • Molecular formula C32H42N4O4, molecular weight 546.70.

Catalog No. 50-002-32944 Supplier Medchemexpress LLC Supplier No. HY12026125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GB-88 is an oral, selective non-peptide antagonist of protease-activated receptor 2 (PAR2) that inhibits PAR2-activated intracellular Ca2+ release with an IC50 of 2 μM. It is supplied as a research reagent, typically provided in milligram-scale packs and commonly dissolved in DMSO for assays.

  • Oral, selective non-peptide PAR2 antagonist.
  • Inhibits PAR2-activated intracellular Ca2+ release (IC50 = 2 μM).
  • High purity suitable for pharmacology studies (98.7%).
  • Supplied in milligram pack sizes for research use.
  • Soluble in DMSO for assay preparation.
  • Molecular formula C32H42N4O4, molecular weight 546.70.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.