Learn More
WELCH MATERIALS INC Welch MaterialsTM Ghost-buster Column, 4.6×50mm
Catalog No. 501963643
Encompass_Preferred
Welch Ghost-Buster Column can effectively remove impurities with low polarity and thus prevents the interference from all kinds of ghost peaks. It is installed between gradient mixer and injector, which helps remove not only the impurities in mobile phase, but impurities in mixer and pipelines as well. Dimension: 4.6×50mm, suitable for ID 4.6mm HPLC column, Max. Pressure: 40 Mpa.
Catalog No. 50-196-3643 Supplier WELCH MATERIALS INC Supplier No. 0610031000
Welch Ghost-Buster Column can effectively remove impurities with low polarity and thus prevents the interference from all kinds of ghost peaks. It is installed between gradient mixer and injector, which helps remove not only the impurities in mobile phase, but impurities in mixer and pipelines as well. Dimension: 4.6×50mm, suitable for ID 4.6mm HPLC column, Max. Pressure: 40 Mpa.