Learn More
WELCH MATERIALS INC Ghost-buster Column Kit, 4.6×50 mm, with 4 peek fittings, 2 peek tubes
Supplier:  WELCH MATERIALS INC GBKIT01
Encompass_Preferred
Welch Ghost-Buster Column can effectively remove impurities with low polarity and thus prevents the interference from all kinds of ghost peaks. It is installed between gradient mixer and injector, which helps remove not only the impurities in mobile phase, but impurities in mixer and pipelines as well. Dimension: 4.6×50mm, suitable for ID 4.6mm HPLC column, Max. Pressure: 40 Mpa.
Catalog No. 50-209-3116