Learn More
Gold Biotechnology Inc GB10B Chemically Competent E. coli Cells 5x50 μl
SDP
Catalog No. NC2942819
Encompass_Preferred
GoldBio’s GB10B Chemically Competent E. coli cells are high efficiency cells, suitable for a wide variety of applications such as cloning and sub-cloning. GB10B Chemically Competent E. coli cells have multiple features including the ɸ80lacZ∆M15 marker, which provides α-complementation of the β-galactosidase gene with blue/white screening protocol. These cells also have the mcrA genotypic marker and the mcrBC, mrr deletion, which allows for cloning of DNA that contains methylcytosine and methyladenine.
Catalog No. NC2942819 Supplier Gold Biotechnology Inc Supplier No. CC1005X50
GoldBio’s GB10B Chemically Competent E. coli cells are high efficiency cells, suitable for a wide variety of applications such as cloning and sub-cloning. GB10B Chemically Competent E. coli cells have multiple features including the ɸ80lacZ∆M15 marker, which provides α-complementation of the β-galactosidase gene with blue/white screening protocol. These cells also have the mcrA genotypic marker and the mcrBC, mrr deletion, which allows for cloning of DNA that contains methylcytosine and methyladenine.