Learn More
Gold Biotechnology Inc GB10BTM Competent E coli Cells
SDP
Catalog No. 5000562190
Encompass_Preferred
GoldBio s GB10B Chemically Competent E coli cells are equivalent to DH10B cells GB10B cells are high efficiency and suitable for a wide variety of applications such as cloning and sub-cloning GB10B Chemically Competent E coli cells have multiple features including the 80lacZ M15 marker which provides -complementation of the -galactosidase gene with blue/white screening protocol These cells also have the mcrA genotypic marker and the mcrBC mrr deletion which allows for cloning of DNA that contains methylcytosine and methyladenine
Catalog No. 50-005-62190 Supplier Gold Biotechnology Inc Supplier No. CC10010X50
GoldBio s GB10B Chemically Competent E coli cells are equivalent to DH10B cells GB10B cells are high efficiency and suitable for a wide variety of applications such as cloning and sub-cloning GB10B Chemically Competent E coli cells have multiple features including the 80lacZ M15 marker which provides -complementation of the -galactosidase gene with blue/white screening protocol These cells also have the mcrA genotypic marker and the mcrBC mrr deletion which allows for cloning of DNA that contains methylcytosine and methyladenine