Learn More
Medchemexpress LLC HY-114409 10mg Medchemexpress, GB1107 CAS:1978336-61-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11440910MG
Encompass_Preferred
Medchemexpress, HY-114409 10mg GB1107 CAS:1978336-61-6 GB1107 is a potent, selective, orally active inhibitor of Galectin-3 (Gal-3) with a Kd of 37 nM for human Galectin-3. GB1107 reduces human and mouse lung adenocarcinoma growth and blocks metastasis in the syngeneic model. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9348