Learn More
Medchemexpress LLC GB1107 is a potent, selective, orally active inhibitor, Solid + Solvent 10 mM 1 mL in DMSO ready for reconstitution
SDP
Catalog No. NC2607345
Encompass_Preferred
GB1107 is a potent, selective, orally active inhibitor of Galectin-3 (Gal-3) with a Kd of 37 nM for human Galectin-3. GB1107 reduces human and mouse lung adenocarcinoma growth and blocks metastasis in the syngeneic model.
Catalog No. NC2607345 Supplier Medchemexpress LLC Supplier No. HY1144091ML/DMSO SOLID
GB1107 is a potent, selective, orally active inhibitor of Galectin-3 (Gal-3) with a Kd of 37 nM for human Galectin-3. GB1107 reduces human and mouse lung adenocarcinoma growth and blocks metastasis in the syngeneic model.