Learn More
Medchemexpress LLC GB111-NH2 hydrochloride | 99.3% | 610.14 | 10 MG
SDP
Catalog No. 5000438571
Encompass_Preferred
GB111-NH2 hydrochloride is a cysteine cathepsin inhibitor and can be used for cancer study.
  • It is a cysteine cathepsin inhibitor.
  • It can be used for cancer study.
  • It is for research use only.
  • It has a purity of 99.26%.
  • Its appearance is a white to off-white solid.
  • It should be shipped at room temperature in continental US and may vary elsewhere.
  • Storage conditions are -20°C, sealed storage, away from moisture and light.
  • In solvent, it should be stored at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light).
  • It is soluble in DMSO at 125 mg/mL.
Catalog No. 50-004-38571 Supplier Medchemexpress LLC Supplier No. HY115454A10MG
GB111-NH2 hydrochloride is a cysteine cathepsin inhibitor and can be used for cancer study.
  • It is a cysteine cathepsin inhibitor.
  • It can be used for cancer study.
  • It is for research use only.
  • It has a purity of 99.26%.
  • Its appearance is a white to off-white solid.
  • It should be shipped at room temperature in continental US and may vary elsewhere.
  • Storage conditions are -20°C, sealed storage, away from moisture and light.
  • In solvent, it should be stored at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture and light).
  • It is soluble in DMSO at 125 mg/mL.