Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GB111-NH2 hydrochloride | 99.3% | 610.14 | 100 MG
SDP

Catalog No. 5000438585
Encompass_Preferred

GB111-NH2 hydrochloride is a cysteine cathepsin inhibitor and can be used for cancer study.

  • Functions as a cysteine cathepsin inhibitor
  • Suitable for cancer research
  • Chemical formula C33H40ClN3O6
  • Appears as a solid, white to off-white powder
  • Soluble in DMSO at 125 mg/mL (in vitro)
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline at 5 mg/mL (in vivo)
  • Classified under metabolic enzyme/protease and cathepsin

Catalog No. 50-004-38585 Supplier Medchemexpress LLC Supplier No. HY115454A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GB111-NH2 hydrochloride is a cysteine cathepsin inhibitor and can be used for cancer study.

  • Functions as a cysteine cathepsin inhibitor
  • Suitable for cancer research
  • Chemical formula C33H40ClN3O6
  • Appears as a solid, white to off-white powder
  • Soluble in DMSO at 125 mg/mL (in vitro)
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline at 5 mg/mL (in vivo)
  • Classified under metabolic enzyme/protease and cathepsin

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.