Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GB111-NH2 hydrochloride | 99.3% | 610.14 g·mol⁻1 | C33H40ClN3O6 | 5 MG
SDP

Catalog No. 5000264427
Encompass_Preferred

GB111-NH2 hydrochloride is a research-grade small-molecule cysteine cathepsin inhibitor used in biochemical and cellular studies, notably in cancer research.

  • High reported purity, 99.3%.
  • Molecular formula C33H40ClN3O6; molar mass 610.14 g·mol⁻1.
  • Physical form: solid, white to off-white.
  • Intended for biochemical and cellular studies related to cathepsin activity and cancer research.
  • Available in milligram-scale packages suitable for screening and mechanistic experiments.
  • Store sealed, away from moisture and light; in solvent store at -80°C for six months or -20°C for one month.

Catalog No. 50-002-64427 Supplier Medchemexpress LLC Supplier No. HY115454A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GB111-NH2 hydrochloride is a research-grade small-molecule cysteine cathepsin inhibitor used in biochemical and cellular studies, notably in cancer research.

  • High reported purity, 99.3%.
  • Molecular formula C33H40ClN3O6; molar mass 610.14 g·mol⁻1.
  • Physical form: solid, white to off-white.
  • Intended for biochemical and cellular studies related to cathepsin activity and cancer research.
  • Available in milligram-scale packages suitable for screening and mechanistic experiments.
  • Store sealed, away from moisture and light; in solvent store at -80°C for six months or -20°C for one month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.