Learn More
Gold Biotechnology Inc GB5-alpha Electrocompetent E. coli Cells 5x50 μl
SDP
Catalog No. NC2942859
Encompass_Preferred
GoldBio’s GB5-alpha Electrocompetent E. coli are high efficiency cells, suitable for a wide variety of applications such as cloning and sub-cloning. Plasmid quality and yields are both enhanced in our GB5-alpha cells due to mutations in both endA1 and recA1.
Catalog No. NC2942859 Supplier Gold Biotechnology Inc Supplier No. CC2035X50C
GoldBio’s GB5-alpha Electrocompetent E. coli are high efficiency cells, suitable for a wide variety of applications such as cloning and sub-cloning. Plasmid quality and yields are both enhanced in our GB5-alpha cells due to mutations in both endA1 and recA1.