Learn More
Gold Biotechnology Inc GB5 alphaTM Comp E coli Cells
SDP
Catalog No. 5000562234
Encompass_Preferred
GoldBio s GB5-alpha Chemically Competent E coli cells are equivalent to DH5-alpha cells and are high efficiency cells suitable for transformation in a wide variety of routine applications such as plasmid isolation cloning and subcloning
Catalog No. 50-005-62234 Supplier Gold Biotechnology Inc Supplier No. CC10110X100
GoldBio s GB5-alpha Chemically Competent E coli cells are equivalent to DH5-alpha cells and are high efficiency cells suitable for transformation in a wide variety of routine applications such as plasmid isolation cloning and subcloning