Learn More
Gold Biotechnology Inc GB5 alphaTM Electro E coli
SDP
Catalog No. 5000562302
Encompass_Preferred
GoldBio s GB5-alpha Electrocompetent E coli cells are equivalent to DH5-alpha cells GB5-alpha cells are high efficiency and suitable for a wide variety of applications such as cloning and sub-cloning Plasmid quality and yields are both enhanced in our GB5-alpha cells due to mutations in both end A1 and rec A1
Catalog No. 50-005-62302 Supplier Gold Biotechnology Inc Supplier No. CC20310X100
GoldBio s GB5-alpha Electrocompetent E coli cells are equivalent to DH5-alpha cells GB5-alpha cells are high efficiency and suitable for a wide variety of applications such as cloning and sub-cloning Plasmid quality and yields are both enhanced in our GB5-alpha cells due to mutations in both end A1 and rec A1