Learn More
Biomatik Corporation Gba Recombinant Antibody
Supplier:  Biomatik Corporation RAC07064
Encompass_Preferred
Gba Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to GBA. The Gba Recombinant Antibody has been validated for the following applications: ELISA, WB. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Gba. Applications: ELISA, WB. Target Synonyms: GBA. Accession Number: P04062. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4411