Learn More
Enterprise Technology Solutions GBC 3000L Laminator, 9"W, Black
SDP
Catalog No. 5000739796
Encompass_Preferred
9" laminator warms up in less than 2 minutes to deliver high-quality results in just moments. Intelligent Jam Alert system beeps until the button is pressed to release a jammed pouch to keep you working efficiently. Versatile design is compatible with 3 mil and 5 mil lamination pouches to provide durable protection that remains flexible. Cold option works with pressure-sensitive, self-adhesive lamination pouches. Green ready light and audible ready alert indicate when it's ready for simple operation. Auto Poweroff turns off the power after 30 minutes of inactivity to help you conserve energy consumption.
Catalog No. 50-007-39796 Supplier Enterprise Technology Solutions Supplier No. GBC1703079
9" laminator warms up in less than 2 minutes to deliver high-quality results in just moments. Intelligent Jam Alert system beeps until the button is pressed to release a jammed pouch to keep you working efficiently. Versatile design is compatible with 3 mil and 5 mil lamination pouches to provide durable protection that remains flexible. Cold option works with pressure-sensitive, self-adhesive lamination pouches. Green ready light and audible ready alert indicate when it's ready for simple operation. Auto Poweroff turns off the power after 30 minutes of inactivity to help you conserve energy consumption.