Learn More
Enterprise Technology Solutions GBC Laminating Pouch, 3 Mil, 11-1/4"x17-1/4", 100/BX, Clear
SDP
Catalog No. 5000740610
Encompass_Preferred
HeatSeal LongLife Laminating Pouches include a UV additive to preserve color integrity and prevent yellowing up to 10 times longer than standard pouches. Enhance your important documents with the best in clarity and protection. Pouch size is perfect for menus, and it offers a 3 mil thickness. Standard construction is designed for occasional handling and filing of documents where pouch flexibility is acceptable.
Catalog No. 50-007-40610 Supplier Enterprise Technology Solutions Supplier No. GBC3200720
HeatSeal LongLife Laminating Pouches include a UV additive to preserve color integrity and prevent yellowing up to 10 times longer than standard pouches. Enhance your important documents with the best in clarity and protection. Pouch size is perfect for menus, and it offers a 3 mil thickness. Standard construction is designed for occasional handling and filing of documents where pouch flexibility is acceptable.