Learn More
Enterprise Technology Solutions GBC Laminating Pouches,Bus Card,7mil,3-11/16"x2-3/16",100/BX,CL
SDP
Catalog No. 5000740011
Encompass_Preferred
Sized to perfectly laminate business cards, thermal laminating pouches provide clean, crisp and professional-looking lamination results. Standard, glossy finish delivers brilliantly clear results. UltraClear technology lets text details and image colors stand out for excellent visibility. 7 mil thickness is durable and semi-rigid to protect against wear and tear. Laminating pouches are designed for use with any thermal pouch laminator (sold separately).
Catalog No. 50-007-40011 Supplier Enterprise Technology Solutions Supplier No. GBC3300371
Sized to perfectly laminate business cards, thermal laminating pouches provide clean, crisp and professional-looking lamination results. Standard, glossy finish delivers brilliantly clear results. UltraClear technology lets text details and image colors stand out for excellent visibility. 7 mil thickness is durable and semi-rigid to protect against wear and tear. Laminating pouches are designed for use with any thermal pouch laminator (sold separately).