Learn More
Enterprise Technology Solutions GBC Laminator,120V/60Hz,12.5 Amps,27" Throat,Adjust Temp,Gray
SDP
Catalog No. 5000740659
Encompass_Preferred
High-volume, extra-wide roll system provides high-quality results and laminates up to 27" wide at 10' per minute. Use with up to 3 mil roll film on a 1" core. Infrared heat rollers warm up in 10 minutes and eliminate cold spots for flawless lamination. One-touch keypad with preset temperature and speed controls makes it easy to use. Protective shield provides added safety. Auto standby after 1 hour and auto shutoff after 2 hours of nonuse conserve energy. LED light and audible alert signal 10-minute warm-up is complete. Laminator offers a built-in horizontal trimmer and motorized reverse function.
Catalog No. 50-007-40659 Supplier Enterprise Technology Solutions Supplier No. GBC1710740
High-volume, extra-wide roll system provides high-quality results and laminates up to 27" wide at 10' per minute. Use with up to 3 mil roll film on a 1" core. Infrared heat rollers warm up in 10 minutes and eliminate cold spots for flawless lamination. One-touch keypad with preset temperature and speed controls makes it easy to use. Protective shield provides added safety. Auto standby after 1 hour and auto shutoff after 2 hours of nonuse conserve energy. LED light and audible alert signal 10-minute warm-up is complete. Laminator offers a built-in horizontal trimmer and motorized reverse function.