Learn More
Enterprise Technology Solutions GBC Laminator, 2-Heat Setting, 35-2/5"x21"x13-7/10", Blue/Gray
SDP
Catalog No. 5000739630
Encompass_Preferred
Pinnacle 27 Thermal Roll Laminator features an 8 to 10 minute warm-up time with an LED light and audible alert signal to let you know when warm-up is complete. It laminates at variable speeds of up to 10' per minute. This laminator includes several user-friendly features like a built-in trimmer, auto reverse, temperature readout and footage counter. The Pinnacle 27 has a maximum document width of 27" that's perfect for laminating classroom charts, certificates, artwork and more. It's compatible with Nap I and Nap II roll film up to 3 mil thick. Laminator goes into auto standby mode after 2 hours without use and auto shutoff after 3 hours without use.
Catalog No. 50-007-39630 Supplier Enterprise Technology Solutions Supplier No. GBC1701700
Pinnacle 27 Thermal Roll Laminator features an 8 to 10 minute warm-up time with an LED light and audible alert signal to let you know when warm-up is complete. It laminates at variable speeds of up to 10' per minute. This laminator includes several user-friendly features like a built-in trimmer, auto reverse, temperature readout and footage counter. The Pinnacle 27 has a maximum document width of 27" that's perfect for laminating classroom charts, certificates, artwork and more. It's compatible with Nap I and Nap II roll film up to 3 mil thick. Laminator goes into auto standby mode after 2 hours without use and auto shutoff after 3 hours without use.