Learn More
Enterprise Technology Solutions GBC ProClick Easy Edit Spines for 11 x 8.5 Documents, 32 Rings, 0.31" (45-Sheet) Capacity, Black, 100/Box
SDP
Catalog No. 5000739805
Encompass_Preferred
Make your presentations stand out with these professional, 32-hole binding spines. Editable design lets you quickly add or remove pages without requiring a machine. Sheets lie flat with a 360-degree rotation to allow easy page-turning and photocopying. 45-sheet capacity is perfect for presentations, proposals and other documents that require multiple edits/updates. Binding spines are designed for use with letter-size papers.
Catalog No. 50-007-39805 Supplier Enterprise Technology Solutions Supplier No. GBC2514700
Make your presentations stand out with these professional, 32-hole binding spines. Editable design lets you quickly add or remove pages without requiring a machine. Sheets lie flat with a 360-degree rotation to allow easy page-turning and photocopying. 45-sheet capacity is perfect for presentations, proposals and other documents that require multiple edits/updates. Binding spines are designed for use with letter-size papers.