Learn More
Enterprise Technology Solutions GBC Roll Film, 1-1/2 mil, 27"x500", 2/BX, Clear
SDP
Catalog No. 5000740214
Encompass_Preferred
Polyester-based thermal film provides excellent bonding strength for items with porous surfaces and light ink coverage. Designed for use on GBC Pinnacle 27, Ultima 65 Catena Series laminators and other hot-roll-style laminators most commonly found in educational environments. Film offers a 1.5 mil thickness and a 1" core diameter. Film is not compatible with heat-shoe-type roll laminators.
Catalog No. 50-007-40214 Supplier Enterprise Technology Solutions Supplier No. GBC3126061
Polyester-based thermal film provides excellent bonding strength for items with porous surfaces and light ink coverage. Designed for use on GBC Pinnacle 27, Ultima 65 Catena Series laminators and other hot-roll-style laminators most commonly found in educational environments. Film offers a 1.5 mil thickness and a 1" core diameter. Film is not compatible with heat-shoe-type roll laminators.