Learn More
Enterprise Technology Solutions GBC Roll Film Laminator,12",120 Vac,Adjust Temp,60Hz,Blue/Gray
SDP
Catalog No. 5000739800
Encompass_Preferred
Perfect for educational settings, daycares, churches and small offices, the Ultima 35 desktop roll laminator delivers high-quality results and features EZ Load technology. EZ Load roll film ensures error-free film loading every time. One-touch AutoSpeed automatically adjusts the laminating speed for you, ensuring quality lamination on every project It warms up in less than 2 minutes and notifies you with a LED ready light. It laminates at 36" per minute. EZ Load technology eliminates film-loading mistakes Laminator also features built-in trimmer, reverse function and maximum document width of 12". Heat Shoe Accutherm heating technology promises flawless, consistent results. Laminator shifts to auto standby after 1 hour and shuts off after 2 hours of nonuse. It is compatible with GBC EZ Load blue endcap roll film with 1.7 to 5 mil thickness on roll with a 1" core and maximum width of 12".
Catalog No. 50-007-39800 Supplier Enterprise Technology Solutions Supplier No. GBC1701680
Perfect for educational settings, daycares, churches and small offices, the Ultima 35 desktop roll laminator delivers high-quality results and features EZ Load technology. EZ Load roll film ensures error-free film loading every time. One-touch AutoSpeed automatically adjusts the laminating speed for you, ensuring quality lamination on every project It warms up in less than 2 minutes and notifies you with a LED ready light. It laminates at 36" per minute. EZ Load technology eliminates film-loading mistakes Laminator also features built-in trimmer, reverse function and maximum document width of 12". Heat Shoe Accutherm heating technology promises flawless, consistent results. Laminator shifts to auto standby after 1 hour and shuts off after 2 hours of nonuse. It is compatible with GBC EZ Load blue endcap roll film with 1.7 to 5 mil thickness on roll with a 1" core and maximum width of 12".