Learn More
Enterprise Technology Solutions GBC Roll Laminator,27"W,12' per Min,w/Trimmer,2 Settings,BE/GY
SDP
Catalog No. 5000740033
Encompass_Preferred
Pinnacle 27 Ezload Roll Laminator laminates documents up to 27" wide at 12' per minute. It is ideal for banners and other continuous feed applications. Innovative Accutherm heating system provides consistent heating and allows you to laminate either Nap 1 or NAP II Roll Film. Ezload film-loading technology allows quick and easy film setup. Easy-to-use control panel features a standby switch, footage counter integrated with a temperature-ready indicator, and power-on LED indicators. Laminator is compatible with 1.0 mil to 3.0 mil NAPI (High Melt) or NAPII (Low Melt) Ezload Film. It features a 6 minute warm-up time, auto-shutoff and an auto-shutoff disengaging switch, built-in trimmer in the back of machine, tray table with safety shutoff, attached film idler bar for easy film loading, LED power and ready lights with audible alert. cUL approved.
Catalog No. 50-007-40033 Supplier Enterprise Technology Solutions Supplier No. GBC1701720EZ
Add to Cart
Add to Cart
Pinnacle 27 Ezload Roll Laminator laminates documents up to 27" wide at 12' per minute. It is ideal for banners and other continuous feed applications. Innovative Accutherm heating system provides consistent heating and allows you to laminate either Nap 1 or NAP II Roll Film. Ezload film-loading technology allows quick and easy film setup. Easy-to-use control panel features a standby switch, footage counter integrated with a temperature-ready indicator, and power-on LED indicators. Laminator is compatible with 1.0 mil to 3.0 mil NAPI (High Melt) or NAPII (Low Melt) Ezload Film. It features a 6 minute warm-up time, auto-shutoff and an auto-shutoff disengaging switch, built-in trimmer in the back of machine, tray table with safety shutoff, attached film idler bar for easy film loading, LED power and ready lights with audible alert. cUL approved.