Learn More
Enterprise Technology Solutions GBC Thermal Laminating Pouch, SPD Format, Ltr, 5 mil, 200/PK, CL
SDP
Catalog No. 5000739741
Encompass_Preferred
Thermal laminating pouches feature helpful guides, thickness icons and directional arrows that disappear after laminating to deliver a crystal-clear finish while ensuring outstanding accuracy. Speed pouches are sealed along the long side to let them run through a laminator faster than standard pouches. Alignment guides on the pouch help you to perfectly center your item inside the pouch to easily create consistently even lamination borders. Thickness icons clearly identify the pouch thickness for proper laminator settings and quality finishing. Directional arrows on the pouch serve as a helpful reminder to ensure you always insert the correct edge first into your laminator, reducing misfeeds and jams. Ultraviolet protection prevents fading and discoloration over time to ensure the smooth, glossy surface always displays dynamic text and rich colors. Letter-size thermal pouches are designed for use with any laminator, including GBC Fusion laminators (sold separately).
Catalog No. 50-007-39741 Supplier Enterprise Technology Solutions Supplier No. GBC3740728
Thermal laminating pouches feature helpful guides, thickness icons and directional arrows that disappear after laminating to deliver a crystal-clear finish while ensuring outstanding accuracy. Speed pouches are sealed along the long side to let them run through a laminator faster than standard pouches. Alignment guides on the pouch help you to perfectly center your item inside the pouch to easily create consistently even lamination borders. Thickness icons clearly identify the pouch thickness for proper laminator settings and quality finishing. Directional arrows on the pouch serve as a helpful reminder to ensure you always insert the correct edge first into your laminator, reducing misfeeds and jams. Ultraviolet protection prevents fading and discoloration over time to ensure the smooth, glossy surface always displays dynamic text and rich colors. Letter-size thermal pouches are designed for use with any laminator, including GBC Fusion laminators (sold separately).