Learn More
Enterprise Technology Solutions GBC Thermal Pouch Laminator, 9-2/5"Wx22-4/5"Lx5-9/10"H, Black
SDP
Supplier:  Enterprise Technology Solutions GBC1703098
Encompass_Preferred
Thermal pouch laminator features highly accurate thickness sensors to let you quickly produce smooth, consistent laminations without having to select a thickness. Fast throughput and one-minute warm-up time provide maximum productivity for busy office environments. Built-in jam detection and auto reverse ensure frustration-free performance throughout large or small projects. Plus, its easy-to-use design with advanced features makes this the perfect unit for both novices and high-frequency users. Manual control is also available for cases with unusual media. Thermal laminator is designed for use with 3 mil to 10 mil thermal pouches.
Catalog No. 50-007-40336