Learn More
Enterprise Technology Solutions GBC Zip Spines, 32-Hole, 5/16"Dia, 45-Sheet Cap, 25/BX, Black
SDP
Catalog No. 5000740034
Encompass_Preferred
Make your presentations stand out with these professional, 32-hole binding spines. Editable design lets you quickly add or remove pages without requiring a machine. Sheets lie flat with a 360-degree rotation to allow easy page-turning and photocopying. 45-sheet capacity is perfect for presentations, proposals and other documents that require multiple edits/updates. Binding spines are designed for use with letter-size papers.
Catalog No. 50-007-40034 Supplier Enterprise Technology Solutions Supplier No. GBC2515662
Make your presentations stand out with these professional, 32-hole binding spines. Editable design lets you quickly add or remove pages without requiring a machine. Sheets lie flat with a 360-degree rotation to allow easy page-turning and photocopying. 45-sheet capacity is perfect for presentations, proposals and other documents that require multiple edits/updates. Binding spines are designed for use with letter-size papers.