Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brand Buzz GBG AloeGel Hand Sanitizer

Catalog No. 22355131
Change view
Click to view available options
Volume (English):
18 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
22-355-131 18 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-355-131 Supplier Brand Buzz Supplier No. HCH 32375
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GBG AloeGel provides broad-spectrum antimicrobial activity, killing 99.9% of harmful microorganisms in 15 seconds. Moisturizes and conditions skin with vitamin E and aloe to help prevent drying, cracking, and irritating of skin. Unique formula helps soften your hands, yet leaves no sticky residue.

  • Pump Bottle
  • Capacity: 18 oz.
  • 12/Pk.

Specifications

Product Type Hand Sanitizer
Color Green
Form Gel
Packaging Pump Bottle
Quantity 12 Pk.
Volume (English) 18 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.