Learn More
Adam Equipment GBK 60aM - GBK-M Approved Bench Checkweighing Scales - Capacity: 60lb / 30kg, Readability: 0.01lb / 5g
Catalog No. NC3964891
Encompass_Preferred
he GBK bench/floor scale combines artful design with reliable performance, offering flexible use with or without a pillar and a durable stainless-steel platform. It supports dynamic weighing, checkweighing, parts counting, percentage weighing, and basic weighing through intuitive software and a color-coded keypad. A large backlit display with capacity tracker shows remaining range during tare to prevent overload. Red, amber, and green LEDs provide clear checkweighing indicators, programmable for quick or detailed feedback. The full numeric keypad enables preset tare and adjustable sample sizes, with supervisor lockout for security. A bi-directional RS-232 interface with date/time allows easy data transfer and traceability. With rechargeable battery, AC power, and automatic accumulation, the GBK delivers productivity and value.
Catalog No. NC3964891 Supplier Adam Equipment Supplier No. GBK 60AM
he GBK bench/floor scale combines artful design with reliable performance, offering flexible use with or without a pillar and a durable stainless-steel platform. It supports dynamic weighing, checkweighing, parts counting, percentage weighing, and basic weighing through intuitive software and a color-coded keypad. A large backlit display with capacity tracker shows remaining range during tare to prevent overload. Red, amber, and green LEDs provide clear checkweighing indicators, programmable for quick or detailed feedback. The full numeric keypad enables preset tare and adjustable sample sizes, with supervisor lockout for security. A bi-directional RS-232 interface with date/time allows easy data transfer and traceability. With rechargeable battery, AC power, and automatic accumulation, the GBK delivers productivity and value.