Learn More
Adam Equipment Adam Equipment GBK-M Bench Checkweighing Scale GBK 150aM 150lb / 60kg capacity x 0.02lb / 0.01kg readability
Catalog No. NC2612406
Encompass_Preferred
"Adam Equipments GBK-M Bench Checkweighing Scales are durable and flexible, GBK-M bench checkweighing scales provide a superb value for businesses that need a scale to perform multiple functions. GBK scales consist of a grade 304 stainless steel weighing pan and sealed keypad to protect against dirt and spills, ideal for messy food operations in bakeries or when weighing chemicals in science laboratories. All GBK-M models feature a full numeric keypad to simplify operation, whilst built-in checkweighing lights and audible checkweighing alarms can be pre-configured to a specified weight for performing accurate parts counting tasks. Ideal for busy production warehouses, shipping and distribution environments. Approved GBK-M models are suitable for use in trade settings where goods are sold by weight. Rechargeable battery included for operation almost anywhere."
Catalog No. NC2612406 Supplier Adam Equipment Supplier No. GBK 150AM
"Adam Equipments GBK-M Bench Checkweighing Scales are durable and flexible, GBK-M bench checkweighing scales provide a superb value for businesses that need a scale to perform multiple functions. GBK scales consist of a grade 304 stainless steel weighing pan and sealed keypad to protect against dirt and spills, ideal for messy food operations in bakeries or when weighing chemicals in science laboratories. All GBK-M models feature a full numeric keypad to simplify operation, whilst built-in checkweighing lights and audible checkweighing alarms can be pre-configured to a specified weight for performing accurate parts counting tasks. Ideal for busy production warehouses, shipping and distribution environments. Approved GBK-M models are suitable for use in trade settings where goods are sold by weight. Rechargeable battery included for operation almost anywhere."