Learn More
Medchemexpress LLC HY-111651 10mg Medchemexpress, Gboxin CAS:2101315-36-8 Purity:>98%
SDP
Catalog No. 501968935
Encompass_Preferred
Medchemexpress, HY-111651 10mg Gboxin CAS:2101315-36-8 Gboxin is an oxidative phosphorylation inhibitor that targets glioblastoma. Gboxin inhibits the activity of F0F1 ATP synthase. Antitumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8935 Supplier Medchemexpress LLC Supplier No. HY11165110MG
Medchemexpress, HY-111651 10mg Gboxin CAS:2101315-36-8 Gboxin is an oxidative phosphorylation inhibitor that targets glioblastoma. Gboxin inhibits the activity of F0F1 ATP synthase. Antitumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.