Learn More
Aviva Systems Biology Corp GBP2 Antibody - N-terminal region (ARP54715_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54715_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GBP2. Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-014-63031