Learn More
Cayman Chemical GBR 12783 (hydrochloride), Purity: ≥98%, CAS: 67469-75-4, 10 mg
SDP
Catalog No. 502910548
Encompass_Preferred
A dopamine uptake inhibitor (IC50 = 13 nM in rat striatal synaptosomes); selective for inhibition of dopamine uptake over norepinephrine or 5-HT uptake (IC50s = 230 and 1,100 nM in rat hypothalamic and striatal synaptosomes, respectively); improves learning and memory in the passive avoidance test in rats at 10 mg/kg, i.p., an effect that can be blocked by scopolamine; increases locomotor activity, reduces pentobarbital-induced sleep duration, and decreases immobility time in the forced swim test, in mice; inhibits haloperidol-induced catalepsy in rats at 10 mg/kg
Catalog No. 50-291-0548 Supplier Cayman Chemical Supplier No. 3566210 MG
A dopamine uptake inhibitor (IC50 = 13 nM in rat striatal synaptosomes); selective for inhibition of dopamine uptake over norepinephrine or 5-HT uptake (IC50s = 230 and 1,100 nM in rat hypothalamic and striatal synaptosomes, respectively); improves learning and memory in the passive avoidance test in rats at 10 mg/kg, i.p., an effect that can be blocked by scopolamine; increases locomotor activity, reduces pentobarbital-induced sleep duration, and decreases immobility time in the forced swim test, in mice; inhibits haloperidol-induced catalepsy in rats at 10 mg/kg