Learn More
Abcam GBR 12909 dihydrochloride, Dopamine transport inhibitor, 50MG
Catalog No. 502707430
Encompass_Preferred
MW 523.5 Da, Purity >98%. Potent, selective dopamine reuptake inhibitor (IC₅₀ = 2.32 μM in rat hippocampal slices; IC₅₀ = 6-35 μM in whole cell patch clamp recordings).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7430 Supplier Abcam Supplier No. AB12060750MG
MW 523.5 Da, Purity >98%. Potent, selective dopamine reuptake inhibitor (IC₅₀ = 2.32 μM in rat hippocampal slices; IC₅₀ = 6-35 μM in whole cell patch clamp recordings).
The product is subject to the following: Abcam Restricted Use Statement