Learn More
Cayman Chemical GBR 12909hydrochlorIde 10mg
SDP
Catalog No. 502969838
Encompass_Preferred
A potent inhibitor that blocks dopamine uptake (IC50 = 1-51 nM); effectively inhibits dopamine uptake in vivo, leading to consequent stimulation of dopamine receptors.
Catalog No. 50-296-9838 Supplier Cayman Chemical Supplier No. 1562210 MG
A potent inhibitor that blocks dopamine uptake (IC50 = 1-51 nM); effectively inhibits dopamine uptake in vivo, leading to consequent stimulation of dopamine receptors.