Learn More
Cayman Chemical GBR 12909 (hydrochloride), Purity: ≥98%, CAS: 67469-78-7, 50 mg
SDP
Catalog No. 502972724
Encompass_Preferred
A potent inhibitor that blocks dopamine uptake (IC50 = 1-51 nM); effectively inhibits dopamine uptake in vivo, leading to consequent stimulation of dopamine receptors.
Catalog No. 50-297-2724 Supplier Cayman Chemical Supplier No. 1562250 MG
A potent inhibitor that blocks dopamine uptake (IC50 = 1-51 nM); effectively inhibits dopamine uptake in vivo, leading to consequent stimulation of dopamine receptors.