Learn More
TARGETMOL CHEMICALS INC GBR 12935 dihydrochloride 50MG
SDP
Catalog No. 502876899
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. GBR 12935 dihydrochloride (GBR 12935) can induce the increase of the extracellular levels of dopamine to basal levels. Purity 99.09%
Catalog No. 50-287-6899 Supplier TARGETMOL CHEMICALS INC Supplier No. T187150MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. GBR 12935 dihydrochloride (GBR 12935) can induce the increase of the extracellular levels of dopamine to basal levels. Purity 99.09%