Learn More
Apexbio Technology LLC GBR 12935 dihydrochloride 67469-81-2 100mg
SDP
Catalog No. 5000090028
Encompass_Preferred
GBR 12935 dihydrochloride is a small-molecule inhibitor targeting the dopamine transporter (DAT) It is designed to inhibit DAT-mediated dopamine reuptake thereby modulating dopaminergic signaling and increasing extracellular dopamine levels GBR 12935 dihydrochloride exerts its biological activity primarily through direct blockade of neuronal dopamine uptake at the synaptic cleft Based on these pharmacological properties GBR 12935 dihydrochloride holds research potential in studies of dopaminergic signaling reward pathways addiction behaviors dopamine-related neurodegeneration and dopamine transporter function
Catalog No. 50-000-90028 Supplier Apexbio Technology LLC Supplier No. B4661100
Add to Cart
Add to Cart
GBR 12935 dihydrochloride is a small-molecule inhibitor targeting the dopamine transporter (DAT) It is designed to inhibit DAT-mediated dopamine reuptake thereby modulating dopaminergic signaling and increasing extracellular dopamine levels GBR 12935 dihydrochloride exerts its biological activity primarily through direct blockade of neuronal dopamine uptake at the synaptic cleft Based on these pharmacological properties GBR 12935 dihydrochloride holds research potential in studies of dopaminergic signaling reward pathways addiction behaviors dopamine-related neurodegeneration and dopamine transporter function