Learn More
Cayman Chemical GBR 12935hydrochlorIde 10mg
SDP
Catalog No. 502965794
Encompass_Preferred
An inhibitor of dopamine uptake; binds to the dopamine transporter (Kd = 27.2 nM); selective for the dopamine transporter over the serotonin and norepinephrine transporters (Kds = 940 and 310 nM, respectively); it increases locomotor activity in mice at 10 mg/kg; induces gnawing in mice (ED50 = 17.1 mg/kg)
Catalog No. 50-296-5794 Supplier Cayman Chemical Supplier No. 1770010 MG
An inhibitor of dopamine uptake; binds to the dopamine transporter (Kd = 27.2 nM); selective for the dopamine transporter over the serotonin and norepinephrine transporters (Kds = 940 and 310 nM, respectively); it increases locomotor activity in mice at 10 mg/kg; induces gnawing in mice (ED50 = 17.1 mg/kg)