Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ GBS Detect™
SDP

Catalog No. 23001790 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-790 100
23-001-736 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-790 Supplier Hardy Diagnostics™ Supplier No. A300BX
Add to Cart
Add to Cart
This item is not returnable. View return policy

GBS Detect™ is recommended for the isolation and differentiation by enhanced hemolysis of detection of gamma-hemolytic Group B Streptococcus from GBS enrichment broth procedures.

  • Optically clear polystyrene for distortion free viewing
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
  • Packaged in sleeves of 10 plates in a breathable bag that prevents buildup of condensation and excess moisture

Specifications

Color Red
Content And Storage 2°C to 8°C
Detects Non-hemolytic Group B Strep
Format Plate
Manufacturing Origin United States
Product Type Agar Plate
Sterility Sterile
Fill Volume 17 mL
For Use With (Application) Isolation and differentiation by enhanced hemolysis of detection of gamma-hemolytic Group B Streptococcus from GBS enrichment broth procedures
Material Polystyrene
No. per Pack 100
Packaging Type Sleeves, Plates
Quantity 100 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Streptococcus species
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.