Learn More
Aviva Systems Biology Corp Gbx1 Antibody - N-terminal region (ARP37213_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37213_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gbx1. It was validated on Western Blot. Gbx1,gastrulation brain homeobox 1,Gbx-1; Gbx1,homeobox protein GBX-1,homeobox protein GBX-1,OTTMUSP00000028788,gastrulation and brain-specific homeobox protein 1The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Gbx-1ApplicationWB. Species reactivity: Dog, Mouse, Pig, Rat
Catalog No. 50-284-596