Learn More
Aviva Systems Biology Corp GBX2 Antibody - middle region (ARP32552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GBX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GBX2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54651