Learn More
ABclonal Technology GBX2 Rabbit pAb
Supplier: ABclonal Technology A147331000UL
Enables sequence-specific double-stranded DNA binding activity. Predicted to be involved in regulation of nervous system development and regulation of transcription by RNA polymerase II. Predicted to act upstream of or within several processes, including branching involved in blood vessel morphogenesis, nervous system development, and neural crest cell migration. Predicted to be part of chromatin. Predicted to be active in nucleus.